The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003880.3 | NP_003871.1 | 1 - 2 | 90 | 71 |
NR_125354.1 | 1 - 2 | 90 | 71 | ||
GenBank mRNA | BC012028.1 | - | 1 - 2 | 90 | 71 |