Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
Chromosome Location
Chr.7: 43608452 - 43729541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135580.1 7 - 8 828 79
XM_017012418.1 XP_016867907.1 6 - 7 718 79
XM_017012430.1 XP_016867919.1 7 - 8 820 79
GenBank mRNA AK023053.1 - 7 - 8 760 79

View Full Product Details