The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006712021.3 | XP_006712084.1 | 2 - 3 | 120 | 92 |
XM_017004150.1 | XP_016859639.1 | 2 - 3 | 120 | 92 | |
XM_017004152.1 | XP_016859641.1 | 1 - 2 | 74 | 92 |