The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014399.3 | NP_055214.1 | 5 - 6 | 789 | 63 |
GenBank mRNA | AF100759.1 | - | 5 - 6 | 709 | 63 |
AF120265.1 | - | 5 - 6 | 615 | 63 | |
AK075475.1 | - | 5 - 6 | 733 | 63 | |
AK093487.1 | - | 5 - 6 | 750 | 63 | |
AK316595.1 | - | 5 - 6 | 732 | 63 | |
AY358995.1 | - | 5 - 6 | 720 | 63 | |
BC033863.1 | - | 5 - 6 | 641 | 63 | |
CR542107.1 | - | 5 - 6 | 547 | 63 |