The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159642.1 | NP_001153114.1 | 8 - 9 | 1363 | 104 |
NM_138278.3 | NP_612122.2 | 8 - 9 | 1111 | 104 | |
XM_017000420.1 | XP_016855909.1 | 9 - 10 | 1158 | 104 | |
GenBank mRNA | AF193056.1 | - | 8 - 9 | 1309 | 104 |
AK092595.1 | - | 8 - 9 | 1063 | 104 | |
AY033000.1 | - | 8 - 9 | 1095 | 104 | |
AY078983.1 | - | 5 - 6 | 692 | 104 | |
AY078984.1 | - | 6 - 7 | 846 | 104 | |
BC027868.1 | - | 8 - 9 | 1088 | 104 | |
BC074779.2 | - | 5 - 6 | 733 | 104 | |
BC074780.2 | - | 5 - 6 | 733 | 104 | |
CU451236.1 | - | 4 - 5 | 453 | 104 |