Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
Chromosome Location
Chr.15: 49125274 - 49155657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143887.1 NP_001137359.1 10 - 11 1141 167
NM_004236.3 NP_004227.1 10 - 11 1120 167
GenBank mRNA AB209799.1 - 10 - 11 1067 167
AF084260.1 - 10 - 11 1056 167
AF100762.1 - 10 - 11 1046 167
AF120268.1 - 10 - 11 1031 167
AF212227.1 - 10 - 11 1067 167
AK222590.1 - 10 - 11 1044 167
AK295598.1 - 8 - 9 875 167
AK312643.1 - 10 - 11 1056 167
BC012629.1 - 10 - 11 1057 167
CR542063.1 - 10 - 11 1041 167

View Full Product Details