Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
Chromosome Location
Chr.15: 49125274 - 49155657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143887.1 NP_001137359.1 2 - 3 245 83
NM_004236.3 NP_004227.1 2 - 3 245 83
GenBank mRNA AB209799.1 - 2 - 3 171 83
AF084260.1 - 2 - 3 181 83
AF100762.1 - 2 - 3 171 83
AF120268.1 - 2 - 3 157 83
AF212227.1 - 2 - 3 171 83
AK222590.1 - 2 - 3 169 83
AK312643.1 - 2 - 3 181 83
BC012629.1 - 2 - 3 182 83
CR542063.1 - 2 - 3 166 83
DB059510.1 - 2 - 3 249 83
L40388.1 - 2 - 3 157 83

View Full Product Details