Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
Chromosome Location
Chr.15: 49125274 - 49155657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143887.1 NP_001137359.1 7 - 8 809 68
NM_004236.3 NP_004227.1 7 - 8 788 68
GenBank mRNA AB209799.1 - 7 - 8 735 68
AF084260.1 - 7 - 8 724 68
AF100762.1 - 7 - 8 714 68
AF120268.1 - 7 - 8 700 68
AF212227.1 - 7 - 8 735 68
AK222590.1 - 7 - 8 712 68
AK295598.1 - 5 - 6 543 68
AK312643.1 - 7 - 8 724 68
BC012629.1 - 7 - 8 725 68
CR542063.1 - 7 - 8 709 68

View Full Product Details