The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018590.4 | NP_061060.3 | 6 - 7 | 1630 | 79 |
NR_135077.1 | 7 - 8 | 1791 | 79 | ||
GenBank mRNA | AB079252.1 | - | 6 - 7 | 1561 | 79 |
AB090811.1 | - | 6 - 7 | 1589 | 79 | |
AK074474.1 | - | 2 - 3 | 282 | 79 | |
AK125300.1 | - | 6 - 7 | 1542 | 79 | |
BC030268.1 | - | 6 - 7 | 1589 | 79 | |
BX640967.1 | - | 7 - 8 | 1687 | 79 | |
BX641073.1 | - | 2 - 3 | 1687 | 79 | |
KU178704.1 | - | 5 - 6 | 1254 | 79 |