The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319654.1 | NP_001306583.1 | 2 - 3 | 1127 | 95 |
NM_001319656.1 | NP_001306585.1 | 2 - 3 | 1127 | 95 | |
NM_018590.4 | NP_061060.3 | 3 - 4 | 1250 | 95 | |
NR_135077.1 | 3 - 4 | 1250 | 95 | ||
XM_011539938.1 | XP_011538240.1 | 3 - 4 | 1250 | 95 | |
XM_017016388.1 | XP_016871877.1 | 3 - 4 | 1250 | 95 | |
XM_017016389.1 | XP_016871878.1 | 3 - 4 | 1250 | 95 | |
XM_017016390.1 | XP_016871879.1 | 3 - 4 | 1250 | 95 | |
GenBank mRNA | AB079252.1 | - | 3 - 4 | 1181 | 95 |
AB090811.1 | - | 3 - 4 | 1209 | 95 | |
AK125300.1 | - | 3 - 4 | 1162 | 95 | |
BC030268.1 | - | 3 - 4 | 1209 | 95 | |
BX640967.1 | - | 3 - 4 | 1189 | 95 | |
CT003455.1 | - | 2 - 3 | 381 | 95 | |
KU178704.1 | - | 2 - 3 | 874 | 95 | |
KU178705.1 | - | 2 - 3 | 874 | 95 | |
KU178706.1 | - | 2 - 3 | 874 | 95 |