The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319654.1 | NP_001306583.1 | 3 - 4 | 1235 | 134 |
NM_018590.4 | NP_061060.3 | 4 - 5 | 1358 | 134 | |
XM_011539938.1 | XP_011538240.1 | 4 - 5 | 1358 | 134 | |
XM_017016388.1 | XP_016871877.1 | 4 - 5 | 1358 | 134 | |
GenBank mRNA | AB079252.1 | - | 4 - 5 | 1289 | 134 |
AB090811.1 | - | 4 - 5 | 1317 | 134 | |
AK125300.1 | - | 4 - 5 | 1270 | 134 | |
BC030268.1 | - | 4 - 5 | 1317 | 134 | |
KU178704.1 | - | 3 - 4 | 982 | 134 | |
KU178705.1 | - | 3 - 4 | 982 | 134 |