Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CISD1
Entrez Gene ID
55847
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
C10orf70, MDS029, ZCD1, mitoNEET
Chromosome Location
Chr.10: 58269102 - 58290553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018464.4 NP_060934.1 1 - 1 121 67
GenBank mRNA AF220049.1 - 1 - 1 59 67
BC005962.1 - 1 - 1 97 67
BC007043.1 - 1 - 1 53 67
BC008474.1 - 1 - 1 58 67
BC059168.1 - 1 - 1 68 67
DA417986.1 - 1 - 1 121 67

View Full Product Details