Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DACT1
Entrez Gene ID
51339
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3
Chromosome Location
Chr.14: 58634068 - 58648321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016651.5 NP_057735.2 3 - 4 797 90
XM_006720167.3 XP_006720230.1 3 - 4 734 90

View Full Product Details