The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003924.3 | NP_003915.2 | 1 - 2 | 602 | 135 |
GenBank mRNA | BC017199.2 | - | 1 - 2 | 338 | 135 |
D82344.1 | - | 1 - 2 | 602 | 135 |