The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323553.1 | NP_001310482.1 | 21 - 22 | 2604 | 86 |
NM_001323554.1 | NP_001310483.1 | 21 - 22 | 3543 | 86 | |
NM_001323555.1 | NP_001310484.1 | 21 - 22 | 3612 | 86 | |
NM_001323556.1 | NP_001310485.1 | 21 - 22 | 2385 | 86 | |
NM_001323557.1 | NP_001310486.1 | 21 - 22 | 2454 | 86 | |
NM_001323558.1 | NP_001310487.1 | 22 - 23 | 2586 | 86 | |
NM_015137.5 | NP_055952.2 | 21 - 22 | 2535 | 86 | |
NR_136615.1 | 22 - 23 | 2834 | 86 | ||
NR_136616.1 | 22 - 23 | 2765 | 86 | ||
GenBank mRNA | AK294485.1 | - | 15 - 16 | 1674 | 86 |
AK302799.1 | - | 12 - 13 | 1215 | 86 | |
BC047323.1 | - | 5 - 6 | 444 | 86 | |
BC071611.1 | - | 21 - 22 | 2512 | 86 | |
BC131486.1 | - | 17 - 18 | 1870 | 86 | |
BC152442.1 | - | 21 - 22 | 2503 | 86 | |
BG392245.1 | - | - | 487 | 86 | |
BQ424552.1 | - | 1 - 2 | 34 | 86 | |
BX648595.1 | - | 8 - 9 | 774 | 86 | |
CR749243.1 | - | 21 - 22 | 2345 | 86 | |
CX165980.1 | - | 3 - 4 | 397 | 86 | |
D63477.1 | - | 21 - 22 | 2503 | 86 |