The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138329.2 | NP_612202.2 | 4 - 5 | 2100 | 90 |
XM_017017252.1 | XP_016872741.1 | 4 - 5 | 2305 | 90 | |
XM_017017253.1 | XP_016872742.1 | 1 - 2 | 1784 | 90 | |
GenBank mRNA | AF479748.1 | - | 4 - 5 | 2100 | 90 |
AY154461.1 | - | 4 - 5 | 2100 | 90 |