Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL3
Entrez Gene ID
26031
Gene Name
oxysterol binding protein like 3
Gene Aliases
ORP-3, ORP3, OSBP3
Chromosome Location
Chr.7: 24796537 - 24980218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015550.3 NP_056365.1 13 - 14 1873 135
NM_145320.2 NP_663160.1 11 - 12 1453 135
NM_145321.2 NP_663161.1 11 - 12 1438 135
NM_145322.2 NP_663162.1 10 - 11 1345 135
NR_104111.1 12 - 13 1546 135
NR_104112.1 11 - 12 1453 135
XM_005249698.3 XP_005249755.1 13 - 14 1593 135
XM_006715681.3 XP_006715744.1 12 - 13 1767 135
XM_006715682.3 XP_006715745.1 12 - 13 1771 135
XM_006715683.3 XP_006715746.1 11 - 12 1678 135
XM_006715684.3 XP_006715747.1 12 - 13 1530 135
XM_011515258.2 XP_011513560.1 13 - 14 1670 135
XM_011515259.2 XP_011513561.1 14 - 15 2167 135
XM_017011948.1 XP_016867437.1 15 - 16 2030 135
XM_017011949.1 XP_016867438.1 13 - 14 2059 135
GenBank mRNA AB014604.2 - 13 - 14 1810 135
AF392444.1 - 13 - 14 1782 135
AF491781.1 - 12 - 13 1500 135
AF491782.1 - 11 - 12 1407 135
AF491783.1 - 11 - 12 1392 135
AF491784.1 - 10 - 11 1299 135
AF491785.1 - 12 - 13 1500 135
AF491786.1 - 11 - 12 1407 135
AF515639.1 - 5 - 6 552 135
AF515640.1 - 4 - 5 459 135
AL832293.1 - 10 - 11 1109 135
AY008372.1 - 13 - 14 1703 135
BC017731.1 - 13 - 14 1802 135

View Full Product Details