Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL3
Entrez Gene ID
26031
Gene Name
oxysterol binding protein like 3
Gene Aliases
ORP-3, ORP3, OSBP3
Chromosome Location
Chr.7: 24796537 - 24980218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015550.3 NP_056365.1 21 - 22 2921 69
NM_145320.2 NP_663160.1 19 - 20 2501 69
NM_145321.2 NP_663161.1 19 - 20 2486 69
NM_145322.2 NP_663162.1 18 - 19 2393 69
NR_104111.1 20 - 21 2993 69
NR_104112.1 19 - 20 2900 69
XM_005249698.3 XP_005249755.1 21 - 22 2641 69
XM_006715681.3 XP_006715744.1 20 - 21 2815 69
XM_006715682.3 XP_006715745.1 20 - 21 2819 69
XM_006715683.3 XP_006715746.1 19 - 20 2726 69
XM_006715684.3 XP_006715747.1 20 - 21 2578 69
XM_011515258.2 XP_011513560.1 21 - 22 2718 69
XM_011515259.2 XP_011513561.1 22 - 23 3215 69
XM_017011948.1 XP_016867437.1 23 - 24 3078 69
XM_017011949.1 XP_016867438.1 21 - 22 3107 69
GenBank mRNA AB014604.2 - 21 - 22 2858 69
AF392444.1 - 21 - 22 2830 69
AF491781.1 - 20 - 21 2548 69
AF491782.1 - 19 - 20 2455 69
AF491783.1 - 19 - 20 2440 69
AF491784.1 - 18 - 19 2347 69
AF491785.1 - 20 - 21 2947 69
AF491786.1 - 19 - 20 2854 69
AL832293.1 - 18 - 19 2157 69
AY008372.1 - 21 - 22 2751 69
BC017731.1 - 21 - 22 2850 69

View Full Product Details