Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL3
Entrez Gene ID
26031
Gene Name
oxysterol binding protein like 3
Gene Aliases
ORP-3, ORP3, OSBP3
Chromosome Location
Chr.7: 24796537 - 24980218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015550.3 NP_056365.1 22 - 23 3038 80
NM_145320.2 NP_663160.1 20 - 21 2618 80
NM_145321.2 NP_663161.1 20 - 21 2603 80
NM_145322.2 NP_663162.1 19 - 20 2510 80
NR_104111.1 21 - 22 3110 80
NR_104112.1 20 - 21 3017 80
XM_005249698.3 XP_005249755.1 22 - 23 2758 80
XM_006715681.3 XP_006715744.1 21 - 22 2932 80
XM_006715682.3 XP_006715745.1 21 - 22 2936 80
XM_006715683.3 XP_006715746.1 20 - 21 2843 80
XM_006715684.3 XP_006715747.1 21 - 22 2695 80
XM_011515258.2 XP_011513560.1 22 - 23 2835 80
XM_011515259.2 XP_011513561.1 23 - 24 3332 80
XM_017011948.1 XP_016867437.1 24 - 25 3195 80
XM_017011949.1 XP_016867438.1 22 - 23 3224 80
GenBank mRNA AB014604.2 - 22 - 23 2975 80
AF392444.1 - 22 - 23 2947 80
AF491781.1 - 21 - 22 2665 80
AF491782.1 - 20 - 21 2572 80
AF491783.1 - 20 - 21 2557 80
AF491784.1 - 19 - 20 2464 80
AF491785.1 - 21 - 22 3064 80
AF491786.1 - 20 - 21 2971 80
AL832293.1 - 19 - 20 2274 80
AY008372.1 - 22 - 23 2868 80
BC017731.1 - 22 - 23 2967 80

View Full Product Details