The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003791.3 | NP_003782.1 | 20 - 21 | 3241 | 73 |
XM_017023816.1 | XP_016879305.1 | 19 - 20 | 2990 | 73 | |
XM_017023817.1 | XP_016879306.1 | 21 - 22 | 3244 | 73 | |
GenBank mRNA | AK091212.1 | - | 9 - 10 | 1951 | 73 |
AK291773.1 | - | 20 - 21 | 3124 | 73 | |
AK299067.1 | - | 19 - 20 | 2466 | 73 | |
AL133583.1 | - | 5 - 6 | 506 | 73 | |
BC114555.1 | - | 20 - 21 | 3127 | 73 | |
BC114961.1 | - | 20 - 21 | 3127 | 73 | |
D42053.1 | - | 20 - 21 | 3204 | 73 |