Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBTPS1
Entrez Gene ID
8720
Gene Name
membrane bound transcription factor peptidase, site 1
Gene Aliases
PCSK8, S1P, SKI-1
Chromosome Location
Chr.16: 84053761 - 84116943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003791.3 NP_003782.1 6 - 7 1384 88
XM_017023816.1 XP_016879305.1 6 - 7 1285 88
XM_017023817.1 XP_016879306.1 6 - 7 1285 88
GenBank mRNA AK291773.1 - 6 - 7 1267 88
AK299067.1 - 5 - 6 609 88
BC114555.1 - 6 - 7 1270 88
BC114961.1 - 6 - 7 1270 88
D42053.1 - 6 - 7 1347 88

View Full Product Details