The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173536.3 | NP_775807.2 | 2 - 3 | 400 | 143 |
GenBank mRNA | AK122845.1 | - | 2 - 3 | 416 | 143 |
AK289806.1 | - | 2 - 3 | 398 | 143 | |
AK299884.1 | - | 2 - 3 | 400 | 143 | |
BC031087.1 | - | 2 - 3 | 336 | 143 | |
BX648937.1 | - | 2 - 3 | 401 | 143 | |
CR933628.1 | - | 2 - 3 | 334 | 143 |