The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003160.2 | NP_003151.2 | 2 - 3 | 157 | 85 |
GenBank mRNA | AK301886.1 | - | 1 - 2 | 733 | 85 |