The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014331.3 | NP_055146.1 | 3 - 4 | 804 | 85 |
XM_011531802.2 | XP_011530104.1 | 3 - 4 | 873 | 85 | |
GenBank mRNA | AB026891.1 | - | 3 - 4 | 759 | 85 |
AB040875.1 | - | 3 - 4 | 659 | 85 | |
AF200708.1 | - | 3 - 4 | 769 | 85 | |
AF252872.1 | - | 3 - 4 | 755 | 85 | |
AJ277882.1 | - | 3 - 4 | 524 | 85 | |
AK290359.1 | - | 3 - 4 | 804 | 85 | |
AK314855.1 | - | 3 - 4 | 698 | 85 | |
BC012087.1 | - | 3 - 4 | 684 | 85 |