Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
Chromosome Location
Chr.9: 35073838 - 35080016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004629.1 NP_004620.1 8 - 9 1567 81
GenBank mRNA AJ007669.1 - 8 - 9 1567 81
AK293427.1 - 9 - 10 1257 81
AK311348.1 - 7 - 8 1041 81
AK312987.1 - 8 - 9 1492 81
BC000032.2 - 8 - 9 1213 81
BC011623.2 - 8 - 9 1340 81
BT009813.1 - 8 - 9 1075 81
KU178053.1 - 8 - 9 1075 81
KU178055.1 - 8 - 9 1075 81
KU178056.1 - 5 - 6 649 81
U70310.1 - 8 - 9 1209 81

View Full Product Details