The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 12 - 13 | 1607 | 140 |
NM_014963.2 | NP_055778.2 | 15 - 16 | 1939 | 140 | |
XM_011527804.2 | XP_011526106.1 | 15 - 16 | 1912 | 140 | |
GenBank mRNA | AB023180.1 | - | 15 - 16 | 1916 | 140 |
AK074102.1 | - | 11 - 12 | 1399 | 140 | |
AK125139.1 | - | 12 - 13 | 1554 | 140 | |
AK292407.1 | - | 15 - 16 | 1939 | 140 | |
AK296849.1 | - | 12 - 13 | 1554 | 140 | |
AK300981.1 | - | 16 - 17 | 1879 | 140 | |
AK309167.1 | - | 12 - 13 | 1554 | 140 | |
BG332816.1 | - | 1 - 2 | 124 | 140 |