The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 23 - 24 | 2940 | 66 |
NM_014963.2 | NP_055778.2 | 26 - 27 | 3272 | 66 | |
XM_011527804.2 | XP_011526106.1 | 26 - 27 | 3245 | 66 | |
GenBank mRNA | AB023180.1 | - | 26 - 27 | 3249 | 66 |
AK074102.1 | - | 22 - 23 | 2732 | 66 | |
AK125139.1 | - | 23 - 24 | 2887 | 66 | |
AK292407.1 | - | 26 - 27 | 3272 | 66 | |
BC106021.1 | - | 3 - 4 | 344 | 66 |