The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002644.3 | NP_002635.2 | 5 - 6 | 1566 | 64 |
XM_011509629.1 | XP_011507931.1 | 5 - 6 | 1477 | 64 | |
GenBank mRNA | AF272149.2 | - | 5 - 6 | 1562 | 64 |
BC110494.2 | - | 5 - 6 | 1463 | 64 | |
BC110495.2 | - | 5 - 6 | 1463 | 64 | |
CR749533.1 | - | 6 - 7 | 1639 | 64 | |
M24559.1 | - | 3 - 4 | 1169 | 64 | |
S62403.1 | - | 5 - 6 | 1538 | 64 | |
X73079.1 | - | 5 - 6 | 1562 | 64 |