The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002644.3 | NP_002635.2 | 7 - 8 | 2070 | 59 |
XM_011509629.1 | XP_011507931.1 | 7 - 8 | 1981 | 59 | |
GenBank mRNA | AF272149.2 | - | 7 - 8 | 2066 | 59 |
BC110494.2 | - | 7 - 8 | 1967 | 59 | |
BC110495.2 | - | 7 - 8 | 1967 | 59 | |
CR749533.1 | - | 8 - 9 | 2143 | 59 | |
M24559.1 | - | 5 - 6 | 1673 | 59 | |
S62403.1 | - | 7 - 8 | 2042 | 59 | |
X73079.1 | - | 7 - 8 | 2066 | 59 |