The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004461.2 | NP_004452.1 | 3 - 4 | 427 | 141 |
GenBank mRNA | AF042347.1 | - | 3 - 4 | 379 | 141 |
AK092093.1 | - | 3 - 4 | 394 | 141 | |
AK130195.1 | - | 3 - 4 | 398 | 141 | |
AK304587.1 | - | 3 - 4 | 399 | 141 | |
BC006495.2 | - | 3 - 4 | 382 | 141 | |
BC043565.1 | - | 3 - 4 | 377 | 141 | |
BT007198.1 | - | 3 - 4 | 379 | 141 | |
CR456740.1 | - | 3 - 4 | 379 | 141 | |
D84471.1 | - | 3 - 4 | 382 | 141 | |
U07424.1 | - | 3 - 4 | 391 | 141 |