The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109.4 | NP_001100.3 | 14 - 15 | 1654 | 73 |
NM_001164489.1 | NP_001157961.1 | 14 - 15 | 1654 | 73 | |
NM_001164490.1 | NP_001157962.1 | 13 - 14 | 1537 | 73 | |
XM_011539117.2 | XP_011537419.2 | 15 - 16 | 1835 | 73 | |
XM_017015465.1 | XP_016870954.1 | 15 - 16 | 1835 | 73 | |
XM_017015466.1 | XP_016870955.1 | 15 - 16 | 1837 | 73 | |
GenBank mRNA | AK301147.1 | - | 13 - 14 | 1502 | 73 |
AK312834.1 | - | 14 - 15 | 1619 | 73 | |
BC064500.1 | - | 14 - 15 | 1601 | 73 | |
BC115404.1 | - | 14 - 15 | 1573 | 73 | |
BI908229.1 | - | 2 - 3 | 73 | ||
D26579.1 | - | 14 - 15 | 1577 | 73 |