The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 21 - 22 | 3291 | 88 |
NM_000642.2 | NP_000633.2 | 21 - 22 | 3213 | 88 | |
NM_000643.2 | NP_000634.2 | 21 - 22 | 3011 | 88 | |
NM_000644.2 | NP_000635.2 | 21 - 22 | 2951 | 88 | |
NM_000645.2 | NP_000636.2 | 19 - 20 | 3024 | 88 | |
NM_000646.2 | NP_000637.2 | 21 - 22 | 3024 | 88 | |
XM_005270557.2 | XP_005270614.1 | 21 - 22 | 2906 | 88 | |
XM_017000501.1 | XP_016855990.1 | 9 - 10 | 1140 | 88 | |
GenBank mRNA | AB208867.1 | - | 20 - 21 | 2817 | 88 |
AB621804.1 | - | 21 - 22 | 3161 | 88 | |
BC078663.1 | - | 21 - 22 | 2933 | 88 | |
M85168.1 | - | 19 - 20 | 3026 | 88 | |
U84007.1 | - | 21 - 22 | 3213 | 88 | |
U84008.1 | - | 21 - 22 | 2951 | 88 | |
U84009.1 | - | 21 - 22 | 3011 | 88 | |
U84010.1 | - | 21 - 22 | 3291 | 88 | |
U84011.1 | - | 21 - 22 | 3024 | 88 |