The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 32 - 33 | 4830 | 148 |
NM_000642.2 | NP_000633.2 | 32 - 33 | 4752 | 148 | |
NM_000643.2 | NP_000634.2 | 32 - 33 | 4550 | 148 | |
NM_000644.2 | NP_000635.2 | 32 - 33 | 4490 | 148 | |
NM_000645.2 | NP_000636.2 | 30 - 31 | 4563 | 148 | |
NM_000646.2 | NP_000637.2 | 32 - 33 | 4563 | 148 | |
XM_005270557.2 | XP_005270614.1 | 32 - 33 | 4445 | 148 | |
XM_017000501.1 | XP_016855990.1 | 20 - 21 | 2679 | 148 | |
GenBank mRNA | AB208867.1 | - | 31 - 32 | 4356 | 148 |
BC078663.1 | - | 32 - 33 | 4472 | 148 | |
M85168.1 | - | 30 - 31 | 4565 | 148 | |
U84007.1 | - | 32 - 33 | 4752 | 148 | |
U84008.1 | - | 32 - 33 | 4490 | 148 | |
U84009.1 | - | 32 - 33 | 4550 | 148 | |
U84010.1 | - | 32 - 33 | 4830 | 148 | |
U84011.1 | - | 32 - 33 | 4563 | 148 |