The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000373.3 | NP_000364.1 | 3 - 4 | 1082 | 85 |
NR_033434.1 | 2 - 3 | 928 | 85 | ||
NR_033437.1 | 4 - 5 | 1181 | 85 | ||
GenBank mRNA | AB062285.1 | - | 3 - 4 | 1004 | 85 |
AK291306.1 | - | 3 - 4 | 998 | 85 | |
AL080099.2 | - | 2 - 3 | 861 | 85 | |
BC000364.2 | - | 3 - 4 | 999 | 85 | |
BC007511.2 | - | 3 - 4 | 999 | 85 | |
CR456787.1 | - | 3 - 4 | 976 | 85 | |
D86227.1 | - | 3 - 4 | 995 | 85 | |
D86228.1 | - | 3 - 4 | 995 | 85 | |
D86230.1 | - | 3 - 4 | 995 | 85 | |
EU921886.1 | - | 3 - 4 | 996 | 85 | |
EU921887.1 | - | 2 - 3 | 842 | 85 | |
EU921888.1 | - | 2 - 3 | 842 | 85 | |
EU921889.1 | - | 3 - 4 | 941 | 85 | |
EU921891.1 | - | 4 - 5 | 1095 | 85 | |
EU921893.1 | - | 3 - 4 | 649 | 85 | |
EU921894.1 | - | 3 - 4 | 996 | 85 | |
J03626.1 | - | 3 - 4 | 1080 | 85 | |
KU178375.1 | - | 3 - 4 | 976 | 85 | |
KU178376.1 | - | 4 - 5 | 1075 | 85 | |
M36661.1 | - | 3 - 4 | 942 | 85 |