Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
Chromosome Location
Chr.10: 90912100 - 90921275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014391.2 NP_055206.2 1 - 2 272 65
GenBank mRNA AU139209.1 - 1 - 2 274 65
AY903446.1 - 1 - 2 41 65
BC005191.1 - 1 - 2 85 65
BC018667.1 - 1 - 2 88 65
X83703.1 - 1 - 2 273 65

View Full Product Details