The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014391.2 | NP_055206.2 | 2 - 3 | 451 | 167 |
GenBank mRNA | AK290960.1 | - | 2 - 3 | 263 | 167 |
AU139209.1 | - | 2 - 3 | 453 | 167 | |
AY903446.1 | - | 2 - 3 | 220 | 167 | |
BC018667.1 | - | 2 - 3 | 267 | 167 |