Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
Chromosome Location
Chr.9: 127815012 - 127854773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000118.3 NP_000109.1 1 - 2 484 148
NM_001114753.2 NP_001108225.1 1 - 2 484 148
GenBank mRNA AK222669.1 - 1 - 2 397 148
AK290389.1 - 1 - 2 368 148
BC014271.2 - 1 - 2 218 148
BC020391.1 - 1 - 2 231 148
DA943458.1 - 1 - 2 479 148
DB079257.1 - 1 - 2 484 148
X72012.1 - 1 - 2 347 148

View Full Product Details