Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRUB2
Entrez Gene ID
26995
Gene Name
TruB pseudouridine synthase family member 2
Gene Aliases
CLONE24922
Chromosome Location
Chr.9: 128309117 - 128322418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015679.1 NP_056494.1 2 - 3 256 96
GenBank mRNA AF131848.1 - 2 - 3 268 96
AJ420462.1 - 2 - 3 260 96
AK001956.1 - 2 - 3 256 96
AK301986.1 - 2 - 3 501 96
BC001457.2 - 2 - 3 275 96

View Full Product Details