Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMPX
Entrez Gene ID
23676
Gene Name
small muscle protein, X-linked
Gene Aliases
DFN6, DFNX4
Chromosome Location
Chr.X: 21705972 - 21758160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045617.1 4 - 5 514 104
GenBank mRNA BF126275.1 - 4 - 5 376 104

View Full Product Details