Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
Chromosome Location
Chr.17: 50111309 - 50129882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257359.1 NP_001244288.1 2 - 3 422 62
NM_174920.3 NP_777580.1 2 - 3 422 62
XM_017024323.1 XP_016879812.1 3 - 4 589 62
XM_017024324.1 XP_016879813.1 2 - 3 1277 62
GenBank mRNA AK094209.1 - 2 - 3 353 62
AK225767.1 - 2 - 3 353 62
AK291482.1 - 2 - 3 349 62
AY148482.1 - 2 - 3 343 62
BC136485.1 - 2 - 3 335 62
BC141822.1 - 2 - 3 362 62
BC142728.1 - 2 - 3 319 62
CN369613.1 - 2 - 3 424 62

View Full Product Details