Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR4
Entrez Gene ID
57088
Gene Name
phospholipid scramblase 4
Gene Aliases
TRA1
Chromosome Location
Chr.3: 146192335 - 146251179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128304.1 NP_001121776.1 9 - 10 1146 90
NM_001128305.1 NP_001121777.1 7 - 8 1108 90
NM_001128306.1 NP_001121778.1 5 - 6 750 90
NM_001177304.1 NP_001170775.1 4 - 5 722 90
NM_020353.2 NP_065086.2 7 - 8 1020 90
XM_005247654.2 XP_005247711.1 8 - 9 1047 90
XM_005247655.2 XP_005247712.1 8 - 9 964 90
XM_011513031.2 XP_011511333.1 7 - 8 906 90
XM_017006921.1 XP_016862410.1 6 - 7 777 90
GenBank mRNA AF087887.1 - 4 - 5 653 90
AF199023.1 - 7 - 8 924 90
AK128442.1 - 4 - 5 622 90
AK290214.1 - 7 - 8 1048 90
AK292815.1 - 9 - 10 1046 90
AK312809.1 - 7 - 8 962 90
AL833760.1 - 7 - 8 934 90
AY550971.1 - 7 - 8 874 90
BC028354.2 - 7 - 8 1020 90

View Full Product Details