The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052941.4 | NP_443173.2 | 1 - 2 | 157 | 101 |
GenBank mRNA | AK131094.1 | - | 1 - 2 | 70 | 101 |
AK312417.1 | - | 1 - 2 | 149 | 101 | |
AL832576.2 | - | 1 - 2 | 87 | 101 | |
BC017889.1 | - | 1 - 2 | 132 | 101 | |
BC070055.1 | - | 1 - 2 | 94 | 101 |