The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278255.1 | NP_001265184.1 | 3 - 4 | 372 | 68 |
NM_017443.4 | NP_059139.3 | 3 - 4 | 402 | 68 | |
NR_027261.1 | 2 - 3 | 536 | 68 | ||
GenBank mRNA | AF226077.1 | - | 2 - 3 | 154 | 68 |
AK074629.1 | - | 2 - 3 | 246 | 68 | |
AK074762.1 | - | 3 - 4 | 325 | 68 | |
AK096050.1 | - | 2 - 3 | 238 | 68 | |
BC003166.1 | - | 2 - 3 | 225 | 68 | |
BC004170.2 | - | 2 - 3 | 225 | 68 | |
DA477404.1 | - | 3 - 4 | 298 | 68 | |
DA908447.1 | - | 3 - 4 | 403 | 68 |