The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052958.2 | NP_443190.2 | 10 - 11 | 1693 | 127 |
XM_011517445.2 | XP_011515747.1 | 10 - 11 | 1656 | 127 | |
XM_011517448.2 | XP_011515750.1 | 9 - 10 | 1536 | 127 | |
GenBank mRNA | AB056652.1 | - | 9 - 10 | 2661 | 127 |
AK094191.1 | - | 4 - 5 | 494 | 127 | |
BC041961.1 | - | 10 - 11 | 1693 | 127 |