The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007278.1 | NP_009209.1 | 1 - 2 | 68 | |
GenBank mRNA | AB030711.1 | - | 1 - 2 | 68 | |
AF044671.1 | - | 1 - 2 | 68 | ||
AF067171.1 | - | 1 - 2 | 68 | ||
AF161586.1 | - | 1 - 2 | 68 | ||
AF183425.1 | - | 1 - 2 | 68 | ||
AL050182.1 | - | - | 231 | 68 | |
BC106748.1 | - | 1 - 2 | 68 | ||
BC106749.1 | - | 1 - 2 | 68 | ||
CR457043.1 | - | 1 - 2 | 68 | ||
CR542235.1 | - | 1 - 2 | 68 | ||
JN663880.1 | - | 1 - 2 | 68 |