The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007278.1 | NP_009209.1 | 3 - 4 | 388 | 78 |
GenBank mRNA | AB030711.1 | - | 3 - 4 | 387 | 78 |
AF044671.1 | - | 3 - 4 | 362 | 78 | |
AF067171.1 | - | 3 - 4 | 388 | 78 | |
AF161586.1 | - | 3 - 4 | 352 | 78 | |
AF183425.1 | - | 3 - 4 | 387 | 78 | |
AK312205.1 | - | 3 - 4 | 383 | 78 | |
AL050182.1 | - | - | 294 | 78 | |
BC106748.1 | - | 3 - 4 | 330 | 78 | |
BC106749.1 | - | 3 - 4 | 330 | 78 | |
CR457043.1 | - | 3 - 4 | 284 | 78 | |
CR542235.1 | - | 3 - 4 | 284 | 78 | |
U65413.1 | - | - | 78 |