Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB8A
Entrez Gene ID
4218
Gene Name
RAB8A, member RAS oncogene family
Gene Aliases
MEL, RAB8
Chromosome Location
Chr.19: 16111680 - 16133635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005370.4 NP_005361.2 5 - 6 642 95
XM_011528020.2 XP_011526322.1 5 - 6 430 95
GenBank mRNA AF498943.1 - 5 - 6 420 95
AK025165.1 - 5 - 6 665 95
AK293676.1 - 5 - 6 425 95
BC002977.1 - 5 - 6 496 95
BT007184.1 - 5 - 6 420 95
CR536583.1 - 5 - 6 420 95
CR542274.1 - 5 - 6 420 95
X56741.1 - 5 - 6 430 95

View Full Product Details