The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267036.1 | NP_001253965.1 | 6 - 7 | 900 | 122 |
NM_001267037.1 | NP_001253966.1 | 5 - 6 | 645 | 122 | |
NM_012244.3 | NP_036376.2 | 8 - 9 | 1861 | 122 | |
NM_182728.2 | NP_877392.1 | 6 - 7 | 799 | 122 | |
NR_049767.1 | 4 - 5 | 672 | 122 | ||
GenBank mRNA | AB037669.1 | - | 8 - 9 | 1824 | 122 |
AF087908.1 | - | 6 - 7 | 754 | 122 | |
AF087911.1 | - | 7 - 8 | 911 | 122 | |
AF135828.1 | - | 8 - 9 | 1336 | 122 | |
AF135829.1 | - | 3 - 4 | 263 | 122 | |
AF135830.1 | - | 4 - 5 | 442 | 122 | |
AF171669.1 | - | 8 - 9 | 1338 | 122 | |
AK223610.1 | - | 8 - 9 | 1860 | 122 | |
AK296702.1 | - | 6 - 7 | 853 | 122 | |
AK300384.1 | - | 5 - 6 | 588 | 122 | |
AK307609.1 | - | 6 - 7 | 733 | 122 | |
AK313465.1 | - | 8 - 9 | 1115 | 122 | |
AL365344.1 | - | 2 - 3 | 176 | 122 | |
AL365345.1 | - | 4 - 5 | 478 | 122 | |
BC036825.1 | - | 4 - 5 | 523 | 122 | |
BC052250.1 | - | 8 - 9 | 1144 | 122 | |
BX248288.1 | - | 6 - 7 | 750 | 122 | |
Y18483.1 | - | 8 - 9 | 1845 | 122 |