The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000780.3 | NP_000771.2 | 5 - 6 | 1273 | 149 |
GenBank mRNA | BC101777.1 | - | 5 - 6 | 1273 | 149 |
BC112184.1 | - | 5 - 6 | 1273 | 149 | |
M93133.1 | - | 5 - 6 | 1269 | 149 | |
X56088.1 | - | 5 - 6 | 1249 | 149 |