The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033449.2 | NP_258260.1 | 10 - 11 | 969 | 77 |
XM_005268524.4 | XP_005268581.1 | 10 - 11 | 996 | 77 | |
XM_006714803.3 | XP_006714866.1 | 9 - 10 | 873 | 77 | |
XM_011537698.2 | XP_011536000.1 | 10 - 11 | 1002 | 77 | |
XM_011537700.2 | XP_011536002.1 | 10 - 11 | 1003 | 77 | |
XM_011537701.2 | XP_011536003.1 | 10 - 11 | 1003 | 77 | |
XM_017010013.1 | XP_016865502.1 | 10 - 11 | 1002 | 77 | |
GenBank mRNA | AK000007.1 | - | 10 - 11 | 953 | 77 |
AY217346.1 | - | 10 - 11 | 969 | 77 | |
AY358478.1 | - | 2 - 3 | 176 | 77 | |
BC047016.1 | - | 9 - 10 | 896 | 77 |